SY3H3D67FB4A Synthonix Corporation

Methyl 5-chloro-3-hydroxythiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 953092-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42718.html

Safety Information

MSDS https://synthonix.com/42718.html
Availability details for
Quantity
  • Availability
  • Add to Cart