SY3H3D67FB4B Synthonix Corporation

2-Methylpyrimidine-4-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 1004-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/42771.html

Safety Information

MSDS https://synthonix.com/42771.html
Availability details for
Quantity
  • Availability
  • Add to Cart