SY3H3D67FB58 Synthonix Corporation

Methyl 6-oxo-1,6-dihydropyridazine-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 89640-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/42790.html

Safety Information

MSDS https://synthonix.com/42790.html
Availability details for
Quantity
  • Availability
  • Add to Cart