SY3H3D67FB5A Synthonix Corporation

Methyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 42821-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42793.html

Safety Information

MSDS https://synthonix.com/42793.html
Availability details for
Quantity
  • Availability
  • Add to Cart