SY3H3D67FB60 Synthonix Corporation

Methyl 5-bromo-1H-pyrrole-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 16420-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42853.html

Safety Information

MSDS https://synthonix.com/42853.html
Availability details for
Quantity
  • Availability
  • Add to Cart