SY3H3D67FB62 Synthonix Corporation

Methyl 2-(2-chlorothiazol-5-yl)acetate

  • MDL number No MDL found
  • Cas Number : 1424352-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42862.html

Safety Information

MSDS https://synthonix.com/42862.html
Availability details for
Quantity
  • Availability
  • Add to Cart