SY3H3D67FB6A Synthonix Corporation

4-Methoxythiophene-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 71050-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/42956.html

Safety Information

MSDS https://synthonix.com/42956.html
Availability details for
Quantity
  • Availability
  • Add to Cart