SY3H3D67FB6F Synthonix Corporation

Methyl 5-bromo-1-methyl-1H-pyrazole-3-carboxylate

  • Cas Number : 1222174-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43035.html

Safety Information

MSDS https://synthonix.com/43035.html
Availability details for
Quantity
  • Availability
  • Add to Cart