SY3H3D67FB75 Synthonix Corporation

3-Methyl-5-nitropyridin-4-amine

  • MDL number No MDL found
  • Cas Number : 18227-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43106.html

Safety Information

MSDS https://synthonix.com/43106.html
Availability details for
Quantity
  • Availability
  • Add to Cart