SY3H3D67FB7C Synthonix Corporation

Methyl 4-Nitro-1H-pyrazole-1-acetate

  • MDL number No MDL found
  • Cas Number : 6715-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43119.html

Safety Information

MSDS https://synthonix.com/43119.html
Availability details for
Quantity
  • Availability
  • Add to Cart