SY3H3D67FB7D Synthonix Corporation

Methyl 1-methyl-4-nitro-1H-imidazole-2-carboxylate

  • Cas Number : 169770-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43121.html

Safety Information

MSDS https://synthonix.com/43121.html
Availability details for
Quantity
  • Availability
  • Add to Cart