SY3H3D67FB83 Synthonix Corporation

5-Methylpyrrole-2-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 1192-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43134.html

Safety Information

MSDS https://synthonix.com/43134.html
Availability details for
Quantity
  • Availability
  • Add to Cart