SY3H3D67FB88 Synthonix Corporation

1-(4-Methyloxazol-2-yl)ethanone

  • Cas Number : 90892-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43141.html

Safety Information

MSDS https://synthonix.com/43141.html
Availability details for
Quantity
  • Availability
  • Add to Cart