SY3H3D67FB8A Synthonix Corporation

1-Methylpyrrole-3-carboxylic Acid

  • MDL number MFCD11111733
  • Cas Number : 36929-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43143.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43143.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart