SY3H3D67FB8C Synthonix Corporation

5-Methyl-1H-pyrrole-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 100047-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43147.html

Safety Information

MSDS https://synthonix.com/43147.html
Availability details for
Quantity
  • Availability
  • Add to Cart