SY3H3D67FB8D Synthonix Corporation

Methyl 4-aminothiophene-2-carboxylate

  • Cas Number : 89499-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43154.html

Safety Information

MSDS https://synthonix.com/43154.html
Availability details for
Quantity
  • Availability
  • Add to Cart