SY3H3D67FB8F Synthonix Corporation

Methyl 5-methylthiazole-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 79247-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/43156.html

Safety Information

MSDS https://synthonix.com/43156.html
Availability details for
Quantity
  • Availability
  • Add to Cart