SY3H3D67FB90 Synthonix Corporation

Methyl 3-methylisoxazole-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 1004-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43162.html

Safety Information

MSDS https://synthonix.com/43162.html
Availability details for
Quantity
  • Availability
  • Add to Cart