SY3H3D67FB92 Synthonix Corporation

5-Methylpyridine-3-sulfonic acid

  • Cas Number : 4808-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43168.html

Safety Information

MSDS https://synthonix.com/43168.html
Availability details for
Quantity
  • Availability
  • Add to Cart