SY3H3D67FB95 Synthonix Corporation

2-(Methylsulfinyl)pyridine

  • Cas Number : 21948-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43172.html

Safety Information

MSDS https://synthonix.com/43172.html
Availability details for
Quantity
  • Availability
  • Add to Cart