SY3H3D67FB99 Synthonix Corporation

4-Methoxy-2-methylpyrimidine

  • Cas Number : 7314-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43205.html

Safety Information

MSDS https://synthonix.com/43205.html
Availability details for
Quantity
  • Availability
  • Add to Cart