SY3H3D67FBA5 Synthonix Corporation

4-Methyl-2-(methylsulfonyl)pyrimidine

  • MDL number No MDL found
  • Cas Number : 77166-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43238.html

Safety Information

MSDS https://synthonix.com/43238.html
Availability details for
Quantity
  • Availability
  • Add to Cart