SY3H3D67FBB3 Synthonix Corporation

(Z)-3-Methylpent-2-en-4-yn-1-ol

  • MDL number No MDL found

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43300.html

Safety Information

MSDS https://synthonix.com/43300.html
Availability details for
Quantity
  • Availability
  • Add to Cart