SY3H3D67FBB7 Synthonix Corporation

1-(Methoxycarbonyl)cyclopropanecarboxylic acid

  • MDL number MFCD06800496
  • Cas Number : 113020-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to yellow or pale-red powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43317.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43317.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart