SY3H3D67FBB9 Synthonix Corporation

2-Methylpyridine-3,4-diamine

  • MDL number No MDL found
  • Cas Number : 15931-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43332.html

Safety Information

MSDS https://synthonix.com/43332.html
Availability details for
Quantity
  • Availability
  • Add to Cart