SY3H3D67FBBE Synthonix Corporation

N2-Methylpyridine-2,5-diamine

  • MDL number No MDL found
  • Cas Number : 28020-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/43342.html

Safety Information

MSDS https://synthonix.com/43342.html
Availability details for
Quantity
  • Availability
  • Add to Cart