SY3H3D67FBBF Synthonix Corporation

1-Methyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole

  • MDL number No MDL found
  • Cas Number : 762233-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/43345.html

Safety Information

MSDS https://synthonix.com/43345.html
Availability details for
Quantity
  • Availability
  • Add to Cart