SY3H3D67FBC0 Synthonix Corporation

6-Methoxypyridine-3,4-diamine

  • MDL number No MDL found
  • Cas Number : 127356-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/43351.html

Safety Information

MSDS https://synthonix.com/43351.html
Availability details for
Quantity
  • Availability
  • Add to Cart