SY3H3D67FBC2 Synthonix Corporation

(3-Methoxypyrazin-2-yl)methanamine

  • MDL number No MDL found
  • Cas Number : 1211580-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43353.html

Safety Information

MSDS https://synthonix.com/43353.html
Availability details for
Quantity
  • Availability
  • Add to Cart