SY3H3D67FBC4 Synthonix Corporation

4-Methoxy-2-methylpyrimidin-5-amine

  • MDL number No MDL found
  • Cas Number : 53135-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/43356.html

Safety Information

MSDS https://synthonix.com/43356.html
Availability details for
Quantity
  • Availability
  • Add to Cart