SY3H3D67FBC9 Synthonix Corporation

Methyl 4-amino-1-methyl-1H-imidazole-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 162085-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43369.html

Safety Information

MSDS https://synthonix.com/43369.html
Availability details for
Quantity
  • Availability
  • Add to Cart