SY3H3D67FBCA Synthonix Corporation

1-(2-Methoxyethyl)-4-nitro-1H-pyrazole

  • MDL number No MDL found
  • Cas Number : 948570-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43371.html

Safety Information

MSDS https://synthonix.com/43371.html
Availability details for
Quantity
  • Availability
  • Add to Cart