SY3H3D67FBCC Synthonix Corporation

(2-(Methylthio)pyrimidin-4-yl)methanamine

  • MDL number No MDL found
  • Cas Number : 1312095-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43374.html

Safety Information

MSDS https://synthonix.com/43374.html
Availability details for
Quantity
  • Availability
  • Add to Cart