SY3H3D67FBD0 Synthonix Corporation

Methyl 3-oxopyrrolidine-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 69079-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43388.html

Safety Information

MSDS https://synthonix.com/43388.html
Availability details for
Quantity
  • Availability
  • Add to Cart