SY3H3D67FBD3 Synthonix Corporation

2-(5-Methylthiazol-4-yl)ethanol

  • MDL number No MDL found
  • Cas Number : 38067-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43392.html

Safety Information

MSDS https://synthonix.com/43392.html
Availability details for
Quantity
  • Availability
  • Add to Cart