SY3H3D67FBD4 Synthonix Corporation

N-Methyl-3-Thiophenemethanamine

  • MDL number No MDL found
  • Cas Number : 210552-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43396.html

Safety Information

MSDS https://synthonix.com/43396.html
Availability details for
Quantity
  • Availability
  • Add to Cart