SY3H3D67FBDA Synthonix Corporation

Methyl 2-oxo-1,2-dihydropyrimidine-5-carboxylate hydrobromide

  • MDL number No MDL found
  • Cas Number : 50628-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43427.html

Safety Information

MSDS https://synthonix.com/43427.html
Availability details for
Quantity
  • Availability
  • Add to Cart