SY3H3D67FBDB Synthonix Corporation

2-Methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide

  • Cas Number : 114086-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43431.html

Safety Information

MSDS https://synthonix.com/43431.html
Availability details for
Quantity
  • Availability
  • Add to Cart