SY3H3D67FBDF Synthonix Corporation

1-Methylpyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD18071558
  • Cas Number : 30727-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to light yellow powder or crysals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43447.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43447.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart