SY3H3D67FBE7 Synthonix Corporation

trans-2-Methylmorpholine-3-carboxylic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 1807882-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/43457.html

Safety Information

MSDS https://synthonix.com/43457.html
Availability details for
Quantity
  • Availability
  • Add to Cart