SY3H3D67FBEE Synthonix Corporation

Methyl 2-aminopentanoate hydrochloride

  • MDL number No MDL found
  • Cas Number : 10047-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43489.html

Safety Information

MSDS https://synthonix.com/43489.html
Availability details for
Quantity
  • Availability
  • Add to Cart