SY3H3D67FBF8 Synthonix Corporation

N-Methylpiperidin-3-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 127294-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43509.html

Safety Information

MSDS https://synthonix.com/43509.html
Availability details for
Quantity
  • Availability
  • Add to Cart