SY3H3D67FBF9 Synthonix Corporation

4-Methylpentan-2-amine hydrochloride

  • MDL number MFCD11101418
  • Cas Number : 71776-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43511.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43511.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart