SY3H3D67FBFA Synthonix Corporation

2-Methyl-3-nitropyridine hydrochloride

  • MDL number No MDL found
  • Cas Number : 63585-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43521.html

Safety Information

MSDS https://synthonix.com/43521.html
Availability details for
Quantity
  • Availability
  • Add to Cart