SY3H3D67FBFF Synthonix Corporation

N2-Methylpyridine-2,5-diamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 117879-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43545.html

Safety Information

MSDS https://synthonix.com/43545.html
Availability details for
Quantity
  • Availability
  • Add to Cart