SY3H3D67FC0A Synthonix Corporation

4-Methoxy-o-phenylenediamine

  • MDL number MFCD00047837
  • Cas Number : 102-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Red-brown to black solid or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43632.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43632.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart