SY3H3D67FC0D Synthonix Corporation

2-Methoxy-5-methylpyridin-4-amine

  • MDL number No MDL found
  • Cas Number : 1260663-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43637.html

Safety Information

MSDS https://synthonix.com/43637.html
Availability details for
Quantity
  • Availability
  • Add to Cart