SY3H3D67FC0E Synthonix Corporation

2-Methoxy-6-(methylamino)pyridine

  • MDL number MFCD00191557
  • Cas Number : 88569-83-9
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/43638.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43638.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart