SY3H3D67FC13 Synthonix Corporation

2-(N-Methylamino)-3-hydroxymethylpyridine

  • Cas Number : 32399-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43647.html

Safety Information

MSDS https://synthonix.com/43647.html
Availability details for
Quantity
  • Availability
  • Add to Cart