SY3H3D67FC15 Synthonix Corporation

Methyl 3-ethyl-1H-pyrazole-5-carboxylate

  • MDL number MFCD08236689
  • Cas Number : 834869-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43659.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43659.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart